Gene Transcripts
- Gene Symbol
- ZNF195
- Entrez Gene ID
- 7748
- Gene Name
- zinc finger protein 195
- Gene Aliases
- HRF1, ZNFP104
- Chromosome Location
- Chr. 11: 3357927 - 3379222 on Build GRCh38
- UniGene ID
- Hs.386294
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB209664.1 |
- |
1 |
10400 |
|
AF003540.1 |
- |
|
2346 |
|
AL833722.1 |
- |
4 |
2180 |
|
View Full Product Details