Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012471.2 | NP_036603.1 | 4 | 1988 | |
XM_017029774.1 | XP_016885263.1 | 5 | 2278 | ||
GenBank mRNA | AB209258.1 | - | 4 | 1402 | |
AF054568.1 | - | 4 | 1987 | ||
BC137271.1 | - | 4 | 1828 | 9021268 | |
BC137274.1 | - | 4 | 1828 | 9021271 |