Gene Transcripts

Gene Symbol
TUFT1
Entrez Gene ID
7286
Gene Name
tuftelin 1
Gene Aliases
Chromosome Location
Chr. 1: 151540294 - 151583583 on Build GRCh38
UniGene ID
Hs.489922
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001126337.1 NP_001119809.1 5, 6 475
NM_001301317.1 NP_001288246.1 7, 8 685
NM_020127.2 NP_064512.1 6, 7 550
XM_011509961.2 XP_011508263.1 4, 5 384
XM_017002223.1 XP_016857712.1 6, 7 626
XM_017002224.1 XP_016857713.1 5, 6 747
GenBank mRNA AF254260.1 - 6, 7 550
AK075001.1 - 5, 6 472
AK223161.1 - 6, 7 546
AK298066.1 - 7, 8 685
AK299925.1 - 5, 6 463
AK315414.1 - 6, 7 546
AL136917.1 - 6, 7 538
AY358610.1 - 5, 6 469
BC002933.1 - 5, 6 443 3956873
BC008301.1 - 6, 7 530 3506004

View Full Product Details