Gene Transcripts

Gene Symbol
TNS1
Entrez Gene ID
7145
Gene Name
tensin 1
Gene Aliases
MST091, MST122, MST127, MSTP091, MSTP122, MSTP127, MXRA6, PPP1R155, TNS
Chromosome Location
Chr. 2: 217799789 - 217978853 on Build GRCh38
UniGene ID
Hs.471381
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308022.1 NP_001294951.1 8 545
NM_001308023.1 NP_001294952.1 8 545
NM_022648.5 NP_072174.3 9 754
XM_011511712.1 XP_011510014.1 5 410
XM_011511713.1 XP_011510015.1 5 410
XM_011511715.1 XP_011510017.1 5 410
XM_011511723.2 XP_011510025.1 9 761
XM_017004801.1 XP_016860290.1 5 410
XM_017004802.1 XP_016860291.1 5 410
XM_017004803.1 XP_016860292.1 5 410
XM_017004804.1 XP_016860293.1 5 410
XM_017004805.1 XP_016860294.1 5 410
XM_017004806.1 XP_016860295.1 5 410
XM_017004807.1 XP_016860296.1 5 410
XM_017004808.1 XP_016860297.1 5 410
XM_017004809.1 XP_016860298.1 9 751
XM_017004810.1 XP_016860299.1 9 634
XM_017004811.1 XP_016860300.1 9 761
XM_017004812.1 XP_016860301.1 8 711
GenBank mRNA AK001785.2 - 9 540
AK300345.1 - 9 751
AK309785.1 - 11 854
AK310169.1 - 6 576
BC051304.1 - 6 621
BC071905.1 - 6 621
BC116187.1 - 7 518 40074822
BC116188.1 - 7 518 40074825
BC126910.1 - 7 518
BC140942.1 - 9 686 9056754

View Full Product Details