Gene Transcripts

Gene Symbol
KRT76
Entrez Gene ID
51350
Gene Name
keratin 76
Gene Aliases
HUMCYT2A, KRT2B, KRT2P
Chromosome Location
Chr. 12: 52768155 - 52777345 on Build GRCh38
UniGene ID
Hs.654392
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015848.4 NP_056932.2 9 1972
GenBank mRNA AK299390.1 - 9 1972
M99063.1 - 9 1949

View Full Product Details