Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016125.3 | NP_057209.3 | 5, 6 | 947 | |
GenBank mRNA | AK098649.1 | - | 3, 4 | 327 | |
AK296197.1 | - | 3, 4 | 750 | ||
BC006971.1 | - | 4, 5 | 824 | ||
BC088365.1 | - | 5, 6 | 910 | 30390062 |