Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001271762.1 | NP_001258691.1 | 11 | 1178 | |
NM_001271763.1 | NP_001258692.1 | 10 | 1153 | ||
NM_017803.4 | NP_060273.1 | 12 | 1258 | ||
GenBank mRNA | AB101210.1 | - | 12 | 922 | |
AK000406.1 | - | 11 | 825 | ||
AK290578.1 | - | 12 | 960 | ||
AK296663.1 | - | 10 | 805 | ||
BC006527.2 | - | 12 | 901 | 2988128 |