Gene Transcripts

Gene Symbol
RSAD1
Entrez Gene ID
55316
Gene Name
radical S-adenosyl methionine domain containing 1
Gene Aliases
Chromosome Location
Chr. 17: 50478800 - 50485980 on Build GRCh38
UniGene ID
Hs.8033
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018346.2 NP_060816.1 9 1850
NR_130911.1 5 1489
GenBank mRNA AK002026.1 - 9 1821
AK022105.1 - 9 1790
AK024628.1 - 4 830
BC005854.2 - 8 1602
BC050538.1 - 9 1781 5214885

View Full Product Details