Gene Transcripts

Gene Symbol
FAM64A
Entrez Gene ID
54478
Gene Name
family with sequence similarity 64 member A
Gene Aliases
CATS, RCS1
Chromosome Location
Chr. 17: 6444305 - 6451065 on Build GRCh38
UniGene ID
Hs.592116
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195228.1 NP_001182157.1 5 1061
NM_019013.2 NP_061886.2 6 1106
XM_017024778.1 XP_016880267.1 4 1435
GenBank mRNA AK001018.1 - 5 1046
AK001353.1 - 5 1021
AK225613.1 - 6 1068
AK225702.1 - 6 1066
BC005004.1 - 5 1028 3533875
BC013966.2 - 6 1052 2988318

View Full Product Details