Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019613.3 | NP_062559.2 | 10 | 1150 | |
XM_005256377.4 | XP_005256434.1 | 9 | 973 | ||
XM_017024862.1 | XP_016880351.1 | 11 | 2487 | ||
XM_017024863.1 | XP_016880352.1 | 12 | 1285 | ||
GenBank mRNA | AF091083.1 | - | 8 | 793 | |
AK297477.1 | - | 10 | 904 | ||
AM182326.1 | - | 10 | 955 | ||
AY691427.1 | - | 8 | 781 | ||
BC000974.2 | - | 10 | 899 | ||
BC007838.1 | - | 10 | 1122 | ||
CR456770.1 | - | 8 | 781 |