Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024320.3 | NP_077296.1 | 2 | 939 | |
XM_005257663.4 | XP_005257720.1 | 2 | 969 | ||
XM_005257664.2 | XP_005257721.1 | 1 | 725 | ||
XM_005257665.1 | XP_005257722.1 | 1 | 725 | ||
XM_005257666.4 | XP_005257723.1 | 3 | 1050 | ||
GenBank mRNA | BC002865.2 | - | 2 | 806 | |
BM831136.1 | - | 1 | 601 |