Gene Transcripts

Gene Symbol
RHBDF2
Entrez Gene ID
79651
Gene Name
rhomboid 5 homolog 2
Gene Aliases
RHBDL5, RHBDL6, TEC, TOC, TOCG, iRhom2
Chromosome Location
Chr. 17: 76470893 - 76501440 on Build GRCh38
UniGene ID
Hs.464157
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005498.3 NP_001005498.2 10, 11 1525
NM_024599.5 NP_078875.4 11, 12 1687
XM_005257669.3 XP_005257726.1 11, 12 2048
XM_005257670.1 XP_005257727.1 11, 12 1557
XM_011525249.2 XP_011523551.1 11, 12 3004
XM_011525250.2 XP_011523552.1 12, 13 1714
XM_011525251.2 XP_011523553.1 11, 12 1531
XM_017025079.1 XP_016880568.1 11, 12 1961
GenBank mRNA AK025994.1 - 6, 7 711
AK074034.1 - 9, 10 1992
AK225684.1 - 6, 7 664
AK292135.1 - 11, 12 1680
AK292166.1 - 10, 11 1480
AL833680.1 - 5, 6 580
AY500247.1 - 10, 11 1376
BC016034.1 - 11, 12 1667 3910543
BC035829.1 - 6, 7 773

View Full Product Details