Gene Transcripts

Gene Symbol
PLEKHH3
Entrez Gene ID
79990
Gene Name
pleckstrin homology, MyTH4 and FERM domain containing H3
Gene Aliases
Chromosome Location
Chr. 17: 42667914 - 42677030 on Build GRCh38
UniGene ID
Hs.632251
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024927.4 NP_079203.3 3 754
NR_073573.1 3 754
NR_073574.1 3 315
XM_017025113.1 XP_016880602.1 4 725
XM_017025114.1 XP_016880603.1 4 725
XM_017025115.1 XP_016880604.1 4 875
XM_017025116.1 XP_016880605.1 3 590
XM_017025117.1 XP_016880606.1 4 725
XM_017025118.1 XP_016880607.1 3 740
GenBank mRNA AB208939.1 - 3 621
AK024672.1 - 3 382
AK096767.1 - 3 315
BC016062.1 - 3 357 4909623
BC052978.1 - 3 675 6473233
HY056649.1 - 3 379

View Full Product Details