Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_032135.3 | NP_115511.3 | 1 | 432 | |
GenBank mRNA | AK124110.1 | - | 1 | 422 | |
AK302615.1 | - | 1 | 386 | ||
BC039878.1 | - | 1 | 391 | 5163160 | |
CR933608.1 | - | 1 | 415 | ||
DB036759.1 | - | 1 | 432 |