Gene Transcripts

Gene Symbol
MFSD5
Entrez Gene ID
84975
Gene Name
major facilitator superfamily domain containing 5
Gene Aliases
hsMOT2
Chromosome Location
Chr. 12: 53251251 - 53254406 on Build GRCh38
UniGene ID
Hs.654660
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170790.1 NP_001164261.1 2 1237
NM_032889.4 NP_116278.3 2 964
XM_005269197.1 XP_005269254.1 3 1492
XM_005269198.4 XP_005269255.1 2 914
GenBank mRNA AK074684.1 - 2 961
AK097576.1 - 2 1170
AK225998.1 - 2 849
AY358604.1 - 2 893
BC007703.2 - 1 780 3948896
BC067795.1 - 2 892 5299341
BX640649.1 - 2 858

View Full Product Details