Gene Transcripts

Gene Symbol
TEKT5
Entrez Gene ID
146279
Gene Name
tektin 5
Gene Aliases
CT149
Chromosome Location
Chr. 16: 10584610 - 10742416 on Build GRCh38
UniGene ID
Hs.143519
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144674.1 NP_653275.1 5 1057
XM_011522383.2 XP_011520685.1 5 1057
XM_011522385.2 XP_011520687.1 5 1057
XM_011522389.1 XP_011520691.1 5 1057
XM_011522390.2 XP_011520692.1 8 1414
XM_017022955.1 XP_016878444.1 5 1057
XM_017022956.1 XP_016878445.1 5 954
XM_017022958.1 XP_016878447.1 2 321
GenBank mRNA AK057433.1 - 5 1057
BC130336.1 - 5 1040 40146367
BC130338.1 - 5 1040 40146369

View Full Product Details