Gene Transcripts

Gene Symbol
C12orf66
Entrez Gene ID
144577
Gene Name
chromosome 12 open reading frame 66
Gene Aliases
Chromosome Location
Chr. 12: 64186312 - 64222296 on Build GRCh38
UniGene ID
Hs.444671
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300941.1 NP_001287870.1 3 1836
NM_152440.4 NP_689653.3 3 1995
GenBank mRNA AK057111.1 - 3 1963
AK129933.1 - 3 1835

View Full Product Details