Gene Transcripts

Gene Symbol
KRT26
Entrez Gene ID
353288
Gene Name
keratin 26
Gene Aliases
CK26, K25, K25IRS2, K26, KRT25B
Chromosome Location
Chr. 17: 40766238 - 40772159 on Build GRCh38
UniGene ID
Hs.592133
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181539.4 NP_853517.2 8 1347
GenBank mRNA AJ564205.2 - 8 1347
BC132951.1 - 8 1338 40146973
BC136896.1 - 8 1338 9020887

View Full Product Details