Gene Transcripts

Gene Symbol
RPS3
Entrez Gene ID
6188
Gene Name
ribosomal protein S3
Gene Aliases
S3
Chromosome Location
Chr. 11: 75399491 - 75422302 on Build GRCh38
UniGene ID
Hs.546286
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005.4 NP_000996.2 5 544
NM_001256802.1 NP_001243731.1 5 544
NM_001260506.1 NP_001247435.1 5 592
NM_001260507.1 NP_001247436.1 4 413
GenBank mRNA AB062288.1 - 5 480
AK130960.1 - 5 466
AK223048.1 - 5 493
AK313051.1 - 5 513
BC003137.1 - 5 493 2906158
BC003577.1 - 5 485
BC013196.1 - 5 485
BC013231.1 - 5 505
BC029981.1 - 4 438
BC034149.1 - 5 517 4665438
BC071917.1 - 5 496 6297012
BC100284.1 - 5 505 6160514
BC150501.1 - 3 319
BE797417.1 - 337
BM928330.1 - 5 574
BQ645330.1 - 552
DW448503.1 - 1 34
S42658.1 - 5 495
U14990.1 - 5 487
U14991.1 - 5 487
U14992.1 - 5 487

View Full Product Details