Gene Transcripts

Gene Symbol
PNN
Entrez Gene ID
5411
Gene Name
pinin, desmosome associated protein
Gene Aliases
DRS, DRSP, SDK3, memA
Chromosome Location
Chr. 14: 39175183 - 39183218 on Build GRCh38
UniGene ID
Hs.409965
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002687.3 NP_002678.2 2, 3 290
GenBank mRNA AK223612.1 - 2, 3 225
AK292579.1 - 2, 3 274
BC062602.1 - 2, 3 201 5491466
CD687183.1 - 2, 3 298
U77718.1 - 2, 3 223
Y09703.1 - 2, 3 201

View Full Product Details