Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002901.2 | NP_002892.1 | 5 | 1019 | |
GenBank mRNA | AK094360.1 | - | 4 | 1109 | |
AK126419.1 | - | 1 | 1350 | ||
AK129791.1 | - | 5 | 1046 | ||
AY423728.1 | - | 3 | 255 | ||
BC010120.2 | - | 5 | 888 | 3836263 | |
D42073.1 | - | 5 | 805 | ||
FJ224346.1 | - | 5 | 805 |