Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002999.3 | NP_002990.2 | 5 | 2045 | |
XM_011528977.2 | XP_011527279.1 | 4 | 1900 | ||
GenBank mRNA | BC030805.1 | - | 5 | 2044 | |
D13292.1 | - | 5 | 2044 | ||
X67016.1 | - | 5 | 2029 |