Gene Transcripts

Gene Symbol
EYA4
Entrez Gene ID
2070
Gene Name
EYA transcriptional coactivator and phosphatase 4
Gene Aliases
CMD1J, DFNA10
Chromosome Location
Chr. 6: 133240340 - 133532120 on Build GRCh38
UniGene ID
Hs.596680
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301012.1 NP_001287941.1 8, 9 966
NM_001301013.1 NP_001287942.1 10, 11 1128
NM_004100.4 NP_004091.3 10, 11 1270
NM_172103.3 NP_742101.2 9, 10 1201
NM_172105.3 NP_742103.1 10, 11 1270
XM_005266851.4 XP_005266908.1 10, 11 977
XM_005266853.4 XP_005266910.1 9, 10 908
XM_017010368.1 XP_016865857.1 11, 12 1497
XM_017010369.1 XP_016865858.1 10, 11 977
XM_017010370.1 XP_016865859.1 9, 10 908
XM_017010371.1 XP_016865860.1 9, 10 893
XM_017010372.1 XP_016865861.1 8, 9 815
XM_017010373.1 XP_016865862.1 8, 9 815
XM_017010374.1 XP_016865863.1 8, 9 1114
XM_017010375.1 XP_016865864.1 8, 9 1088
GenBank mRNA AK295798.1 - 8, 9 1115
AK299378.1 - 10, 11 1042
AK301950.1 - 9, 10 1206
BC014193.2 - 2, 3 109
BC041063.1 - 9, 10 1140 5458911
KU178044.1 - 8, 9 743
KU178045.1 - 7, 8 650
Y17114.1 - 10, 11 1253

View Full Product Details