Gene Transcripts
- Gene Symbol
- PSMD5
- Entrez Gene ID
- 5711
- Gene Name
- proteasome 26S subunit, non-ATPase 5
- Gene Aliases
- S5B
- Chromosome Location
- Chr. 9: 120798318 - 120843021 on Build GRCh38
- UniGene ID
- Hs.193725
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001270427.1 |
NP_001257356.1
|
1, 2 |
288 |
|
NM_005047.3 |
NP_005038.1
|
1, 2 |
288 |
|
XM_011518866.2 |
XP_011517168.1
|
1, 2 |
198 |
|
XM_017014927.1 |
XP_016870416.1
|
1, 2 |
198 |
|
XM_017014928.1 |
XP_016870417.1
|
1, 2 |
198 |
|
GenBank mRNA |
AK303007.1 |
- |
1, 2 |
218 |
|
BC014478.1 |
- |
1, 2 |
177 |
4878087 |
BM835267.1 |
- |
1, 2 |
195 |
|
D31889.1 |
- |
1, 2 |
175 |
|
HY023459.1 |
- |
1, 2 |
289 |
|
S79862.1 |
- |
1, 2 |
179 |
|
View Full Product Details