Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005399.4 | NP_005390.1 | 8 | 3343 | |
NR_103870.1 | 6 | 3055 | |||
NR_103871.1 | 7 | 3176 | |||
XM_011509729.1 | XP_011508031.1 | 9 | 3380 | ||
XM_017001697.1 | XP_016857186.1 | 7 | 3222 | ||
GenBank mRNA | AK023717.1 | - | 1 | 1957 | |
AV660341.1 | - | 1 | 225 | ||
BC053610.1 | - | 8 | 3296 | 6091476 |