Gene Transcripts

Gene Symbol
SOS1
Entrez Gene ID
6654
Gene Name
SOS Ras/Rac guanine nucleotide exchange factor 1
Gene Aliases
GF1, GGF1, GINGF, HGF, NS4
Chromosome Location
Chr. 2: 38981549 - 39124959 on Build GRCh38
UniGene ID
Hs.709893
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005633.3 NP_005624.2 9 1175
XM_005264515.4 XP_005264572.1 9 1404
XM_011533062.2 XP_011531364.1 9 1343
XM_011533064.2 XP_011531366.1 9 1083
XM_011533066.2 XP_011531368.1 2 84
GenBank mRNA AB209140.1 - 2 146
AK290228.1 - 10 1221
AK301960.1 - 10 1321
AL833457.1 - 4 1014
L13857.1 - 9 1134

View Full Product Details