Gene Transcripts

Gene Symbol
SKP2
Entrez Gene ID
6502
Gene Name
S-phase kinase-associated protein 2, E3 ubiquitin protein ligase
Gene Aliases
FBL1, FBXL1, FLB1, p45
Chromosome Location
Chr. 5: 36151989 - 36192792 on Build GRCh38
UniGene ID
Hs.23348
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005983.3 NP_005974.2 5 780
NM_032637.3 NP_116026.1 5 780
XM_011514081.2 XP_011512383.1 5 834
XM_011514082.2 XP_011512384.1 5 834
XM_011514083.2 XP_011512385.1 4 583
XM_017009753.1 XP_016865242.1 4 583
GenBank mRNA AB050979.1 - 4 560
AB050980.1 - 4 560
AB050981.1 - 4 560
AK291255.1 - 5 712
AK295152.1 - 5 654
AY029177.1 - 5 612
BC001441.2 - 5 699 3138709
BC007441.2 - 5 703 2962938

View Full Product Details