Gene Transcripts
- Gene Symbol
- DRG1
- Entrez Gene ID
- 4733
- Gene Name
- developmentally regulated GTP binding protein 1
- Gene Aliases
- NEDD3
- Chromosome Location
- Chr. 22: 31399553 - 31434186 on Build GRCh38
- UniGene ID
- Hs.115242
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004147.3 |
NP_004138.1
|
6, 7 |
845 |
|
GenBank mRNA |
AF078103.1 |
- |
6, 7 |
775 |
|
AJ005940.1 |
- |
6, 7 |
779 |
|
AK098576.1 |
- |
6, 7 |
721 |
|
AK315659.1 |
- |
6, 7 |
803 |
|
AL133085.1 |
- |
1, 2 |
64 |
|
BC019285.1 |
- |
6, 7 |
784 |
2823263 |
BC020803.1 |
- |
6, 7 |
779 |
4250998 |
BT007237.1 |
- |
6, 7 |
714 |
|
CR456488.1 |
- |
6, 7 |
756 |
|
CR542059.1 |
- |
6, 7 |
714 |
|
View Full Product Details