Gene Transcripts

Gene Symbol
SNTB2
Entrez Gene ID
6645
Gene Name
syntrophin beta 2
Gene Aliases
D16S2531E, EST25263, SNT2B2, SNT3, SNTL
Chromosome Location
Chr. 16: 69186133 - 69309052 on Build GRCh38
UniGene ID
Hs.461117
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006750.3 NP_006741.1 2 632
GenBank mRNA AF243385.1 - 2 632
BC048215.1 - 2 617 5805707
U40572.1 - 2 632

View Full Product Details