Gene Transcripts

Gene Symbol
RTN1
Entrez Gene ID
6252
Gene Name
reticulon 1
Gene Aliases
NSP
Chromosome Location
Chr. 14: 59595976 - 59871288 on Build GRCh38
UniGene ID
Hs.368626
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021136.2 NP_066959.1 4 2156
NM_206852.2 NP_996734.1 2 551
XM_017021578.1 XP_016877067.1 2 440
GenBank mRNA AK290534.1 - 2 551
AK297492.1 - 3 458
BC000314.2 - 2 336
BC003003.1 - 2 336
BC090862.1 - 4 2071 6179803
BC111694.1 - 5 1846 40035553
BX449967.2 - 2 539
CR457030.1 - 2 243
L10333.1 - 4 2069
L10334.1 - 2 977
L10335.1 - 2 271

View Full Product Details