Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001136026.2 | NP_001129498.1 | 10 | 1447 | |
NM_001136232.2 | NP_001129704.1 | 9 | 1301 | ||
NM_030673.3 | NP_109598.2 | 10 | 1303 | ||
NM_183352.2 | NP_899195.1 | 9 | 1269 | ||
XM_005265379.2 | XP_005265436.1 | 10 | 1320 | ||
XM_017007019.1 | XP_016862508.1 | 10 | 1357 | ||
XM_017007020.1 | XP_016862509.1 | 9 | 937 | ||
XM_017007021.1 | XP_016862510.1 | 9 | 947 | ||
GenBank mRNA | AB209554.1 | - | 9 | 3443 | |
AF052155.1 | - | 9 | 1301 | ||
AK095629.1 | - | 7 | 3128 | ||
BC006167.1 | - | 6 | 961 | ||
BC091506.1 | - | 9 | 1179 | 30408210 | |
DB564012.1 | - | 1 | 245 | ||
L09260.1 | - | 9 | 1146 |