Gene Transcripts
- Gene Symbol
- MSI1
- Entrez Gene ID
- 4440
- Gene Name
- musashi RNA binding protein 1
- Gene Aliases
- Chromosome Location
- Chr. 12: 120339655 - 120369180 on Build GRCh38
- UniGene ID
- Hs.158311
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002442.3 |
NP_002433.1
|
4, 5 |
349 |
|
XM_006719403.3 |
XP_006719466.1
|
4, 5 |
343 |
|
XM_011538361.2 |
XP_011536663.1
|
4, 5 |
343 |
|
XM_011538362.2 |
XP_011536664.1
|
4, 5 |
343 |
|
XM_011538363.2 |
XP_011536665.1
|
4, 5 |
343 |
|
GenBank mRNA |
AB012851.1 |
- |
4, 5 |
324 |
|
AB072590.1 |
- |
1, 2 |
71 |
|
AB072591.1 |
- |
1, 2 |
71 |
|
AB072592.1 |
- |
1, 2 |
71 |
|
AB073212.1 |
- |
1, 2 |
64 |
|
BX393884.2 |
- |
4, 5 |
349 |
|
View Full Product Details