Gene Transcripts

Gene Symbol
MYO9B
Entrez Gene ID
4650
Gene Name
myosin IXB
Gene Aliases
CELIAC4, MYR5
Chromosome Location
Chr. 19: 17075781 - 17213295 on Build GRCh38
UniGene ID
Hs.123198
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130065.1 NP_001123537.1 7, 8 1489
NM_004145.3 NP_004136.2 7, 8 1489
GenBank mRNA AB209997.1 - 7, 8 1484
AB290184.1 - 7, 8 1484

View Full Product Details