Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005563.3 | NP_005554.1 | 5 | 1376 | |
NM_203399.1 | NP_981944.1 | 5 | 1352 | ||
NM_203401.1 | NP_981946.1 | 5 | 1564 | ||
GenBank mRNA | AK054594.1 | - | 6 | 1272 | |
J04991.1 | - | 5 | 1303 | ||
X53305.1 | - | 5 | 1281 | ||
Z11566.1 | - | 5 | 1291 |