Gene Transcripts

Gene Symbol
FOXO4
Entrez Gene ID
4303
Gene Name
forkhead box O4
Gene Aliases
AFX, AFX1, MLLT7
Chromosome Location
Chr. X: 71096149 - 71103534 on Build GRCh38
UniGene ID
Hs.584654
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170931.1 NP_001164402.1 3 998
NM_005938.3 NP_005929.2 2 1163
XM_011530957.2 XP_011529259.1 2 580
GenBank mRNA AF384029.1 - 3 618
AK300142.1 - 2 1109
BC026735.1 - 1 272
BC106761.2 - 2 889 40025381
X93996.1 - 2 953

View Full Product Details