Gene Transcripts
- Gene Symbol
- PCNT
- Entrez Gene ID
- 5116
- Gene Name
- pericentrin
- Gene Aliases
- KEN, MOPD2, PCN, PCNT2, PCNTB, PCTN2, SCKL4
- Chromosome Location
- Chr. 21: 46324103 - 46445769 on Build GRCh38
- UniGene ID
- Hs.474069
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001315529.1 |
NP_001302458.1
|
9, 10 |
1735 |
|
NM_006031.5 |
NP_006022.3
|
9, 10 |
1573 |
|
XM_005261124.4 |
XP_005261181.1
|
9, 10 |
1592 |
|
XM_011529594.2 |
XP_011527896.1
|
10, 11 |
1673 |
|
XM_017028362.1 |
XP_016883851.1
|
9, 10 |
1592 |
|
XM_017028363.1 |
XP_016883852.1
|
9, 10 |
1761 |
|
GenBank mRNA |
AB007862.3 |
- |
9, 10 |
1736 |
|
AF515282.1 |
- |
9, 10 |
1466 |
|
BC035913.1 |
- |
10, 11 |
1515 |
|
U52962.1 |
- |
9, 10 |
1518 |
|
View Full Product Details