Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001170460.1 | NP_001163931.1 | 14 | 1661 | |
NM_006201.4 | NP_006192.1 | 14 | 1796 | ||
NM_033018.3 | NP_148978.2 | 14 | 1903 | ||
XM_011543920.2 | XP_011542222.1 | 14 | 1673 | ||
XM_011543921.1 | XP_011542223.1 | 15 | 2068 | ||
XM_011543922.1 | XP_011542224.1 | 15 | 2056 | ||
XM_011543923.1 | XP_011542225.1 | 14 | 1986 | ||
XM_011543924.1 | XP_011542226.1 | 14 | 1796 | ||
XM_011543925.2 | XP_011542227.1 | 15 | 1878 | ||
XM_011543926.1 | XP_011542228.1 | 14 | 2839 | ||
XM_011543927.2 | XP_011542229.1 | 15 | 1916 | ||
XM_011543928.1 | XP_011542230.1 | 15 | 2271 | ||
XM_017029569.1 | XP_016885058.1 | 15 | 2068 | ||
XM_017029570.1 | XP_016885059.1 | 15 | 1878 | ||
XM_017029571.1 | XP_016885060.1 | 14 | 2839 | ||
XM_017029572.1 | XP_016885061.1 | 15 | 1916 | ||
XM_017029573.1 | XP_016885062.1 | 15 | 2271 | ||
GenBank mRNA | AB209465.1 | - | 11 | 2033 | |
AK131486.1 | - | 10 | 2578 | ||
AK290145.1 | - | 14 | 1720 | ||
AK301832.1 | - | 14 | 1661 | ||
AK303849.1 | - | 15 | 1676 | ||
BC001048.2 | - | 14 | 1799 | 3504276 | |
BC006190.2 | - | 14 | 1613 | 3830397 | |
BC009852.2 | - | 12 | 1059 | ||
BC015607.2 | - | 14 | 1766 | 4762081 | |
BT006827.1 | - | 13 | 1361 | ||
X66363.1 | - | 14 | 1484 |