Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012328.2 | NP_036460.1 | 2 | 454 | |
GenBank mRNA | AB026908.1 | - | 2 | 287 | |
AF083247.1 | - | 2 | 288 | ||
AF093824.1 | - | 2 | 254 | ||
AK092204.1 | - | 3 | 312 | ||
AL080081.1 | - | 2 | 334 | ||
AY359045.1 | - | 1 | 108 | ||
BC028912.1 | - | 2 | 268 | 4943933 | |
CR533475.1 | - | 1 | 85 | ||
DA202575.1 | - | 2 | 454 |