Gene Transcripts

Gene Symbol
OGN
Entrez Gene ID
4969
Gene Name
osteoglycin
Gene Aliases
OG, OIF, SLRR3A
Chromosome Location
Chr. 9: 92383271 - 92404699 on Build GRCh38
UniGene ID
Hs.109439
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014057.4 NP_054776.1 7 2661
NM_024416.4 NP_077727.3 7 2580
NM_033014.3 NP_148935.1 7 2943
GenBank mRNA AF086912.1 - 7 2508
AF173383.1 - 1 737
AL133114.1 - 3 1926
BC037273.2 - 6 2449 5265880
BC095443.1 - 6 2465 30405707
CB268982.1 - 1 351

View Full Product Details