Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000210.3 | NP_000201.2 | 2 | 526 | |
NM_001079818.2 | NP_001073286.1 | 2 | 526 | ||
NM_001316306.1 | NP_001303235.1 | 2 | 227 | ||
XM_006712510.1 | XP_006712573.1 | 2 | 495 | ||
XM_006712511.1 | XP_006712574.1 | 2 | 495 | ||
XM_017004005.1 | XP_016859494.1 | 2 | 222 | ||
XM_017004006.1 | XP_016859495.1 | 2 | 222 | ||
XM_017004007.1 | XP_016859496.1 | 2 | 222 | ||
XM_017004008.1 | XP_016859497.1 | 2 | 222 | ||
GenBank mRNA | AB208842.1 | - | 2 | 194 | |
AK296496.1 | - | 2 | 163 | ||
AK311695.1 | - | 2 | 227 | ||
BC007002.1 | - | 2 | 492 | ||
BC136455.1 | - | 2 | 400 | 9020444 | |
BC136456.1 | - | 2 | 400 | 9020445 | |
DA760250.1 | - | 2 | 495 | ||
X53586.1 | - | 2 | 438 | ||
X59512.1 | - | 2 | 274 |