Gene Transcripts

Gene Symbol
FLNA
Entrez Gene ID
2316
Gene Name
filamin A
Gene Aliases
ABP-280, ABPX, CSBS, CVD1, FLN, FLN-A, FLN1, FMD, MNS, NHBP, OPD, OPD1, OPD2, XLVD, XMVD
Chromosome Location
Chr. X: 154348529 - 154374638 on Build GRCh38
UniGene ID
Hs.195464
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110556.1 NP_001104026.1 22 3904
NM_001456.3 NP_001447.2 22 3904
XM_011531127.1 XP_011529429.1 22 3899
XM_011531128.1 XP_011529430.1 22 3899
XM_011531129.1 XP_011529431.1 22 3899
XM_011531130.1 XP_011529432.1 22 3899
XM_011531131.1 XP_011529433.1 21 3698
GenBank mRNA AB191259.1 - 21 3627
AB191260.1 - 21 3632
AB371574.1 - 21 3632
AB371575.1 - 21 3627
AB371576.1 - 21 3632
AB371577.1 - 21 3627
AB371578.1 - 21 3627
AB371579.1 - 21 3627
AB593010.1 - 21 3767
AK074048.1 - 1 104
AK090427.2 - 21 3693
GU727643.1 - 22 3904
X53416.1 - 22 3827

View Full Product Details