Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001168298.1 | NP_001161770.1 | 4 | 2132 | |
NM_001557.3 | NP_001548.1 | 3 | 2340 | ||
XM_005246530.3 | XP_005246587.1 | 4 | 2112 | ||
XM_017003990.1 | XP_016859479.1 | 4 | 2092 | ||
XM_017003991.1 | XP_016859480.1 | 2 | 2003 | ||
XM_017003992.1 | XP_016859481.1 | 2 | 2273 | ||
GenBank mRNA | BC037961.1 | - | 3 | 2294 | |
L19593.1 | - | 3 | 2327 |