Gene Transcripts

Gene Symbol
ACKR1
Entrez Gene ID
2532
Gene Name
atypical chemokine receptor 1 (Duffy blood group)
Gene Aliases
CCBP1, CD234, DARC, Dfy, FY, GPD, GpFy, WBCQ1
Chromosome Location
Chr. 1: 159204013 - 159206500 on Build GRCh38
UniGene ID
Hs.153381
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001122951.2 NP_001116423.1 1 1212
NM_002036.3 NP_002027.2 2 1978
GenBank mRNA AK291593.1 - 2 1978
BC017817.1 - 2 1183 4696425
BX537430.1 - 3 1875
U01839.1 - 1 1212

View Full Product Details