Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002221.3 | NP_002212.3 | 6, 7 | 2889 | |
GenBank mRNA | AJ242780.1 | - | 5, 6 | 2606 | |
AK289700.1 | - | 5, 6 | 1335 | ||
AK313801.1 | - | 5, 6 | 1354 | ||
AY513281.1 | - | 6, 7 | 1130 | ||
X57206.1 | - | 5, 6 | 1228 | ||
Y18024.2 | - | 5, 6 | 2549 |