Gene Transcripts

Gene Symbol
TMSB4Y
Entrez Gene ID
9087
Gene Name
thymosin beta 4, Y-linked
Gene Aliases
TB4Y
Chromosome Location
Chr. Y: 13703567 - 13706024 on Build GRCh38
UniGene ID
Hs.159201
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004202.2 NP_004193.1 2 1107
GenBank mRNA AF000989.1 - 2 1105
BC022482.1 - 2 370

View Full Product Details