Gene Transcripts

Gene Symbol
CYTH3
Entrez Gene ID
9265
Gene Name
cytohesin 3
Gene Aliases
ARNO3, GRP1, PSCD3
Chromosome Location
Chr. 7: 6161779 - 6272611 on Build GRCh38
UniGene ID
Hs.487479
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004227.3 NP_004218.1 13 1791
GenBank mRNA AJ223957.1 - 13 1712
BC008191.1 - 6 1030
BC028717.1 - 13 1827 4811560

View Full Product Details