Gene Transcripts

Gene Symbol
UBE2K
Entrez Gene ID
3093
Gene Name
ubiquitin conjugating enzyme E2 K
Gene Aliases
E2-25K, HIP2, HYPG, LIG, UBC1
Chromosome Location
Chr. 4: 39698044 - 39782792 on Build GRCh38
UniGene ID
Hs.50308
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001111112.1 NP_001104582.1 3, 4 508
NM_001312647.1 NP_001299576.1 3, 4 420
NM_001312648.1 NP_001299577.1 2, 3 414
NM_005339.4 NP_005330.1 3, 4 508
GenBank mRNA AB022435.1 - 3, 4 310
AK093533.1 - 3, 4 329
AK307130.1 - 2, 3 319
AK315524.1 - 3, 4 434
BC022804.1 - 3, 4 384 4893778
BC050600.1 - 3, 4 362 6502901
CX786829.1 - 3, 4 347
DA526283.1 - 3, 4 508
U58522.1 - 3, 4 227

View Full Product Details