Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019111.4 | NP_061984.2 | 5 | 1139 | |
GenBank mRNA | AK297032.1 | - | 5 | 790 | |
AK301134.1 | - | 5 | 1036 | ||
AK301354.1 | - | 4 | 780 | ||
AK304429.1 | - | 4 | 507 | ||
AK310161.1 | - | 4 | 587 | ||
BC032350.1 | - | 5 | 1070 | 5214156 | |
BC071659.1 | - | 5 | 1078 | 5162925 | |
J00194.1 | - | 5 | 1057 | ||
K01171.1 | - | 5 | 1044 | ||
V00523.1 | - | 4 | 939 | ||
V00528.1 | - | 2 | 362 |