Gene Transcripts

Gene Symbol
IFI6
Entrez Gene ID
2537
Gene Name
interferon alpha inducible protein 6
Gene Aliases
6-16, FAM14C, G1P3, IFI-6-16, IFI616
Chromosome Location
Chr. 1: 27666061 - 27672213 on Build GRCh38
UniGene ID
Hs.523847
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002038.3 NP_002029.3 5 487
NM_022872.2 NP_075010.1 5 499
NM_022873.2 NP_075011.1 5 511
GenBank mRNA AK024814.1 - 5 467
AK314488.1 - 5 478
BC011601.2 - 5 438 3140035
BC015603.2 - 5 440 4654295
BT006850.1 - 4 364
X02492.1 - 5 433

View Full Product Details