Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000397.3 | NP_000388.2 | 6 | 656 | |
GenBank mRNA | AK289753.1 | - | 6 | 641 | |
AK301491.1 | - | 5 | 518 | ||
AK304033.1 | - | 5 | 470 | ||
AK312929.1 | - | 6 | 616 | ||
BC032720.1 | - | 6 | 642 | 5505170 | |
KU178009.1 | - | 6 | 595 | ||
X04011.1 | - | 6 | 610 |